Lucifer yellow CH dilithium salt 100 mM in Wasser fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulphonic acid dilithium salt, LY CH lithium salt

80026
BTIU80026EA 347 EUR
BTIU80026
Lucifer yellow CH dilithium salt 100 mM in Wasser fluoreszierender Farbstoff
Lucifer yellow CH dilithium salt

Lucifer Yellow CH lithium salt (LY CH lithium salt) is a widely used polar molecular tracer for studying neuronal morphology. The fluorescent molecule contains a carbohydrazide that allow the molecule to be aldehyde-fixable. The lithium salt form of Lucifer Yellow is in an aqueous solution format that is ready for microinjection.


  • λEx/λ Em=428/536 nm
  • Yellow solution. Store at room temperature and protect from light.

Formel: C₁₃H₉N₅O₉S₂Li₂
Molecular Weight: 457,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006922
CAS-Nummer: 67769-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a37d0e54e33266ea24800068e5ef261e5fd81e0&vid=4a37d0e54e33266ea24800068e5ef261e5fd81e0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR