Endotoxin standard, PyroDetect
Lieferant: Merck
1.44161.0001
BTSM1.44161.0001EA 1020 EUR
BTSM1.44161.0001
Endotoxin standard, PyroDetect
Standards Biologische Standards
PyroDetect is used to perform an endotoxin standard curve using different concentrations of endotoxin in dilution.
  • One of the two optional standards
  • Standard curve required to quantify the amount of pyrogens
  • Determine the limit of detection (LOD) of the PyroDetect System
To perfom the endotoxin standard curve, the PyroDetect Endotoxin Standard or any other International Reference Standard Endotoxin (RSE) can be used.