Rotary evaporator, large scale, Rotavapor® R-250
Lieferant: BUCHI
8500C511 8500C512 8500C521 8500C522 8500C541 8500C542 8500C551 8500C552 8500D511 8500D512 8500D521 8500D522 8500D541 8500D542 8500D551 8500D552 8500G511 8500G512 8500G521 8500G522 8500G541 8500G542 8500G551 8500G552 8501C511 8501C512 8501C521 8501C522 8501C541 8501C542 8501C551 8501C552 8501D511 8501D512 8501D521 8501D522 8501D541 8501D542 8501D551 8501D552 8501G511 8501G512 8501G521 8501G522 8501G541 8501G542 8501G551 8501G552
BUCH8500C511EA 0 EUR
BUCH8500C511 BUCH8500C512 BUCH8500C521 BUCH8500C522 BUCH8500C541 BUCH8500C542 BUCH8500C551 BUCH8500C552 BUCH8500D511 BUCH8500D512 BUCH8500D521 BUCH8500D522 BUCH8500D541 BUCH8500D542 BUCH8500D551 BUCH8500D552 BUCH8500G511 BUCH8500G512 BUCH8500G521 BUCH8500G522 BUCH8500G541 BUCH8500G542 BUCH8500G551 BUCH8500G552 BUCH8501C511 BUCH8501C512 BUCH8501C521 BUCH8501C522 BUCH8501C541 BUCH8501C542 BUCH8501C551 BUCH8501C552 BUCH8501D511 BUCH8501D512 BUCH8501D521 BUCH8501D522 BUCH8501D541 BUCH8501D542 BUCH8501D551 BUCH8501D552 BUCH8501G511 BUCH8501G512 BUCH8501G521 BUCH8501G522 BUCH8501G541 BUCH8501G542 BUCH8501G551 BUCH8501G552
Rotary evaporator, large scale, Rotavapor® R-250
Verdampfer Rotationsverdampfer
Batches up to 30 liters can be quickly and safely distilled in the 50-liter evaporating flask of the R-250. The powerful heating bath allows a distillation rate up to 30 liters of acetone per hour.
  • Fast distillation thanks to high heating capacity
  • Intuitive operation; large and easy to read display
  • Easy flask handling due to the unique flask coupling
  • Spring-loaded valves protect the glass
  • Long life time thanks to exclusive use of corrosion-free stainless steel
  • Highly chemical resistant materials
  • Trouble-free operation thanks to robust industrial valves