Pure Essential Chromatography System
Lieferant: BUCHI
11C10700
BUEF11C10700EA 4900 EUR
BUEF11C10700
Pure Essential Chromatography System
Chromatographiesysteme Flash-Chromatographiesysteme
The Pure essential flash chromatography system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
  • 50 bar pump, pressure sensors, and regulators
  • 4 fixed UV wavelengths for most common applications
  • Liquid or solid sample loading
  • Cartridges from 4 to 5000 g or glass columns
  • Enclosed and vented fraction collector bay
  • Wavelength: 254/275/325/365 nm
The small footprint of the instrument allows for placement even in crowded labs, and its robustness makes it highly reliable. Basic but powerful and safe thanks to a 3-piston pump. Expansion possibilities include the addition of a UV detector and fraction collector to the Pure Chromatography C-900 pump.
Intuitive navigation is provided by novel and state-of-the-art software (methods, run data, and general settings). All tubing and parts of the instrument are easily accessible and allow the user to follow and monitor the process flow.
Standards: CE and CSA certified.
Bestellinformation: Fraction racks, collection vessels, cartridge stand and clamp are not included with components or systems to allow customisation and must be ordered separately.