Spigot, stainless steel
Lieferant: BURKLE
0535-0020
BURK0535-0020EA 192.3 EUR
BURK0535-0020
Spigot, stainless steel
Hähne
It is Very sturdy and robust spigot made of stainless steel. The outlet of the spigot turns 360° so it can be adjusted to any required position.
  • Seals PTFE, FKM
  • Self-closing
  • Lockable
  • NW 10 mm