Allschicht-Bohrer
Lieferant: BURKLE
5310-0150
BURK5310-0150EA 1512.75 EUR
BURK5310-0150
Allschicht-Bohrer
Probenehmer Probenehmer für Feststoffe
This sampler with drilling blades makes it easy to take samples of media, even those with high density, and every type of highly compacted powders and granulates.
This special zone collector is suitable for heavy and compact substances – in large containers, mixed containers, big bags in the building materials, chemical, pharmaceutical, food and luxury items industries.