Piston pump
Lieferant: BURKLE
5613-0030
BURK5613-0030EA 37.75 EUR
BURK5613-0030
Piston pump
Pumpen Fass/Transferpumpen
Piston pump is used for watery and petrochemical products.
Pump capacity adjustable from 100, 250, 350 and 500 ml per stroke. The piston pump is attached to the container which is to be filled using a 2" or 3/4" barrel thread. After mounting, the pump can still be rotated around its axis so that containers to be filled can be placed on the barrel lid.