Lever pump, aluminium
Lieferant: BURKLE
5627-1000
BURK5627-1000EA 234.65 EUR
BURK5627-1000
Lever pump, aluminium
Pumpen Fass/Transferpumpen
Lever pump is used for petrochemical substances.
  • With closure device
  • Immersion tube of length 840 mm
Lieferumfang: Supplied with hose kit, hose length 1,5 m, electrically conductive, with outlet elbow and bend protection spring.