Drum pump sets for solvents
Lieferant: BURKLE
5763-0301
BURK5763-0301EA 4353.65 EUR
BURK5763-0301
Drum pump sets for solvents
Pumpen Fass/Transferpumpen
Barrel pump is used for highly flammable hydrocarbons such as ethanol, benzene, butanol, isopropanol, kerosene, methanol and many more.
  • Pump tube made of stainless steel
  • Delivery pump height 14 mWS
Lieferumfang: Supplied with motor 230 V, voltage balancing cable, hose electrically-conductive, (2 m) solvent hose (3/4"), brass pump nozzle, PP barrel screw joint.