Anti-DNA-PK Rabbit Polyclonal Antibody [clone: Y2052]

Lieferant: Bioworld Technology
BS1634
BWRLBS1634EA 262 EUR
BWRLBS1634
Anti-DNA-PK Rabbit Polyclonal Antibody [clone: Y2052]
Antikörper
Synthetic peptide, corresponding to amino acids 2022-2070 of Human DNA-PK.

DNA-PK (Y2052) pAb detects endogenous levels of DNA-PK protein.

Type: Primary
Antigen: DNA-PK
Clonality: Polyclonal
Clone: Y2052
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR