Anti-ACTR-IB Rabbit Polyclonal Antibody [clone: G111]

Lieferant: Bioworld Technology
BS1988
BWRLBS1988EA 262 EUR
BWRLBS1988
Anti-ACTR-IB Rabbit Polyclonal Antibody [clone: G111]
Antikörper
Synthetic peptide, corresponding to amino acids 100-150 of Human ACTR-IB.

ACTR-IB (G111) pAb detects endogenous levels of ACTR-IB protein.

Type: Primary
Antigen: ACTR-IB
Clonality: Polyclonal
Clone: G111
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR