Anti-BMP10 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5628
BWRLBS5628EA 262 EUR
BWRLBS5628
Anti-BMP10 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 282-327 of Human BMP10.

BMP10 pAb detects endogenous levels of BMP10 protein.

Type: Primary
Antigen: BMP10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR