Anti-CXADR Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5638
BWRLBS5638EA 262 EUR
BWRLBS5638
Anti-CXADR Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 48-93 of Human CXADR.

CXADR pAb detects endogenous levels of CXADR protein.

Type: Primary
Antigen: CXADR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR