Anti-RAB27A Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7324
BWRLBS7324EA 341 EUR
BWRLBS7324
Anti-RAB27A Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human RAB27A.

RAB27A pAb detects endogenous levels of RAB27A protein.

Type: Primary
Antigen: RAB27A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR