Anti-MVP Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7339
BWRLBS7339EA 341 EUR
BWRLBS7339
Anti-MVP Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human MVP.

MVP pAb detects endogenous levels of MVP protein.

Type: Primary
Antigen: MVP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR