Anti-CD244 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7344
BWRLBS7344EA 341 EUR
BWRLBS7344
Anti-CD244 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human CD244.

CD244 pAb detects endogenous levels of CD244 protein.

Type: Primary
Antigen: CD244
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR