Anti-GNB2L1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7383
BWRLBS7383EA 341 EUR
BWRLBS7383
Anti-GNB2L1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GNB2L1.

GNB2L1 pAb detects endogenous levels of GNB2L1 protein.

Type: Primary
Antigen: GNB2L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR