Anti-GZMA Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7978
BWRLBS7978EA 341 EUR
BWRLBS7978
Anti-GZMA Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GZMA.

GZMA pAb detects endogenous levels of GZMA protein.

Type: Primary
Antigen: GZMA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR