Anti-STK33 Rabbit Polyclonal Antibody [clone: N108]

Lieferant: Bioworld Technology
BS9197
BWRLBS9197EA 262 EUR
BWRLBS9197
Anti-STK33 Rabbit Polyclonal Antibody [clone: N108]
Antikörper
Synthetic peptide, corresponding to amino acids 72-124 of Human STK33.

STK33 (N108) pAb detects endogenous levels of STK33 protein.

Type: Primary
Antigen: STK33
Clonality: Polyclonal
Clone: N108
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR