Citalopram-D6 hydrobromide solution, certified reference material, Supelco®
Lieferant: Merck
C-090-1ML
C-090-1MLEA
219
EUR
C-090-1ML
Citalopram-D6 hydrobromide solution, certified reference material, Supelco®
Standards
Organische Standards
Citalopram is an antidepressant sold under the trade names Celexa® and Cipramil for the treatment of major depression.
Citalopram is a selective serotonin reuptake inhibitor, a class of drugs that also includes fluoxetine, paroxetine, and sertraline. This internal standard is suitable for quantitation of citalopram levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
Verpackung: Snap-N-Spike®/Snap-N-Shoot®