Human pulmonary microvascular endothelial cells (HPMEC)
Lieferant: PROMOCELL
C-14027
C-14027EA 380 EUR
C-14027
Human pulmonary microvascular endothelial cells (HPMEC)
Zellen
Primary Human Pulmonary Microvascular Endothelial Cells (HPMEC) are isolated from the lung from a single donor. Since lung tissue contains blood and lymphatic capillaries, HPMEC comprise Blood and Lymphatic Microvascular Endothelial Cells. The cells are routinely analysed by immunofluorescent staining: they stain positive for CD31 and von Willebrand factor and negative for smooth muscle alpha-actin.
The lung has a vast endothelial surface area, which is essential for the exchange of gases. HPMEC are most appropriate for studying human lung diseases.
Applications include: Angiogenesis research, oncology, drug uptake studies, cell-cell junction investigation, inflammation studies.
Bestellinformation: Human Pulmonary Microvascular Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.