Human Uterine microvascular endothelial cells (HUtMEC)
Lieferant: PROMOCELL
C-14028
C-14028EA 369 EUR
C-14028
Human Uterine microvascular endothelial cells (HUtMEC)
Zellen
Primary Human Uterine Microvascular Endothelial Cells (HUtMEC) are isolated from the myometrium (middle layer of the uterine wall) from a single donor who is not pre-treated with hormones. The cells are routinely analysed by immunofluorescent staining: they stain positive for CD31 and von Willebrand factor and negative for smooth muscle alpha-actin.
During pregnancy HUtMEC propagate intensely and change their phenotype and expression pattern in response to sex steroids. This includes up-regulation of HUtMEC nitric oxide synthase activity, modulation of adhesion molecule expression, and an increase in endothelial cell survival and angiogenic activity. HUtMEC are involved in medical conditions like uterine fibroma or adenomysis.
Applications include: Angiogenesis research, oncology, drug uptake studies, cell-cell junction investigation, inflammation studies.
Bestellinformation: Human Human Uterine Microvascular Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.