Human pulmonary fibroblasts
Lieferant: PROMOCELL
C-14035
C-14035EA 266 EUR
C-14035
Human pulmonary fibroblasts
Zellen
Primary Human Pulmonary Fibroblasts (HPF) are isolated from human lung tissue
  • CD90 positive
  • Cryopreserved at second passage
  • Available from different tissues
This product can be used in the following applications. Wound healing research, Tissue engineering research, Regeneration studies, Toxicological research, Oncology, Epithelial-mesenchymal interaction studies.
They are often used for studying lung tissue repair, tissue remodeling after injury, or the response after pulmonary inflammation. Fibroblast disorders like pulmonary fibrosis or hypoxia-induced pulmonary hypertension can also be investigated with cultured HPF.
Bestellinformation: Human Pulmonary Fibroblasts can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.