Human primary cells, human bronchial smooth muscle cells (HBSMC)
Lieferant: PROMOCELL
C-14055
C-14055EA 382 EUR
C-14055
Human primary cells, human bronchial smooth muscle cells (HBSMC)
Zellen
Primary Human Bronchial Smooth Muscle Cells (HBSMC) are isolated from the bronchi from single donors and stain positive for smooth muscle α-actin.
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
  • Applications: Organ specific drug testing, cell-extracellular matrix interaction studies, physiological and biochemical studies, oncology, pathological studies (e.g. hypertension)
HBSMC connect the endings of the bronchial cartilage and consequently regulate the airway lumen. Under pathological conditions like asthma, the amount of smooth muscle cells in the bronchi walls is increased leading to smooth muscle hypertrophy or hyperplasia. Hence, HBSMC are suitable for in vitro studies of asthma and other pulmonary diseases.