Human nasal epithelial cells (HNEpC)
Lieferant: PROMOCELL
Primary human nasal epithelial cells (HNEpC) are isolated from normal human nasal mucosa and stain positive for cytokeratin. The epithelium in the nasal cavity cleans, humidifies, and warms inhaled air. In addition, it produces mucus, which binds particles that are subsequently transported to the pharynx by cilia on the epithelial cells. HNEpC are useful for in vitro studies of these processes.
- Cytokeratin positive
- Routinely morphology checked
- Cryopreserved at second passage
This product can be used in the following applications. Drug uptake studies, Toxicological research, Wound healing research, Oncology, Cell differentiation studies, Respiratory disease research.
Lieferumfang: Human Nasal Epithelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.