Human mesenchymal stem cells from bone marrow
Lieferant: PROMOCELL
C-14090
C-14090EA
367
EUR
C-14090
Human mesenchymal stem cells from bone marrow
Zellen
Mesenchymal stem cells are harvested from normal human bone marrow. All cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes, osteoblasts and neuronal cells.
PromoCell's hMSC-BM cells show a verified marker expression profile that complies with ISCT* recommendations, providing well characterised cells.
*ISCT (International Society for Cellular Therapy) Cytotherapy (2006) Vol. 8, No. 4, 315-317