Human mesenchymal stem cells from adipose tissue
Lieferant: PROMOCELL
C-14092
C-14092EA 361 EUR
C-14092
Human mesenchymal stem cells from adipose tissue
Zellen
Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT) are harvested from normal human adipose tissue. The cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes and osteoblasts.
Applications include: Stem cell research, regenerative medicine research, tissue engineering studies, developmental experiments.
PromoCell's hMSC-BM cells show a verified marker expression profile that complies with ISCT* recommendations, providing well characterised cells.
*ISCT (International Society for Cellular Therapy) Cytotherapy (2006) Vol. 8, No. 4, 315-317