Human mononuclear cells from cord blood (hMNC-CB)
Lieferant: PROMOCELL
C-14096
C-14096EA 239 EUR
C-14096
Human mononuclear cells from cord blood (hMNC-CB)
Zellen
Human mononuclear cells from cord blood (hMNC-CB) are isolated from umbilical cord blood, which has a much higher content of primitive progenitor cells compared to adult peripheral blood.
Applications include: Stem cell research, transplantation basic research, hematologic and immunologic studies, oncology research, virology and infection biology.