Skeletal muscle cell differentiation medium
Lieferant: PROMOCELL
C-23061
C-23061EA
210
EUR
C-23061
Skeletal muscle cell differentiation medium
Zellkulturmedien
PromoCell Skeletal Muscle Cell Differentiation Medium is a serum-free medium that induces the fusion of myoblasts to myotubes with typical multinucleated syncytia.
The medium is optimised for primary human cells, but can also be used for murine, rat, canine and rabbit skeletal muscle cells.
Lieferumfang: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix.