Osteoblast mineralisation medium
Lieferant: PROMOCELL
C-27020
C-27020EA 216 EUR
C-27020
Osteoblast mineralisation medium
Zellkulturmedien
PromoCell Osteoblast Mineralisation Medium has been developed for the efficient induction of strong mineralisation in human osteoblast cultures. Extensive extracellular calcium deposits can be observed after 21 days with an easy, one-step protocol.