Preadipocyte differentiation medium
Lieferant: PROMOCELL
C-27437
C-27437EA 340 EUR
C-27437
Preadipocyte differentiation medium
Zellkulturmedien
PromoCell Preadipocyte Differentiation Medium is a serum-free medium which induces the differentiation of preadipocytes into mature adipocytes.
Lieferumfang: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. Kit consists of 1× 500 ml bottle of basal medium and 1× SupplementPack (individual vials with pre-measured supplements).