Mesenchymal stem cell growth medium
Lieferant: PROMOCELL
C-28009
C-28009EA 287 EUR
C-28009
Mesenchymal stem cell growth medium
Zellkulturmedien
PromoCell Mesenchymal Stem Cell Growth Medium supports the expansion of multipotent human mesenchymal stem cells (MSC) without inducing early senescence and differentiation as observed in standard culture media.
Lieferumfang: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix.