Mesenchymal stem cell growth medium DXF
Lieferant: PROMOCELL
PromoCell Mesenchymal Stem Cell Growth Medium DXF is a chemically defined and xeno-free formula, which strongly supports self-renewal and maximum growth of multipotent MSC. It is devoid of all animal-derived components and substances of human origin with human serum albumin as the only exception. Fibronectin-coated plates are necessary in conjunction with this medium.