Mononuclear cell medium
Lieferant: PROMOCELL
C-28030
C-28030EA
291
EUR
C-28030
Mononuclear cell medium
Zellkulturmedien
PromoCell Mononuclear Cell Medium is a very rich short-term maintenance medium which provides utmost cell viability. It is suitable to maintain medium for up to 48 hours before proceeding with your experiments.
Recommended for human mononuclear cells from cord blood (hMNC-CB), human mononuclear cells from peripheral blood (hMNC-PB), CD14+ monocytes.
Lieferumfang: The medium consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix.