Monocyte attachment medium
Lieferant: PROMOCELL
C-28051
C-28051EA
129
EUR
C-28051
Monocyte attachment medium
Zellkulturmedien
PromoCell Monocyte Attachment Medium allows for the efficient adherence selection of monocytes from freshly isolated mononuclear cells (MNC) while maintaining optimal cell health. Thus, the time consuming and costly immunomagnetic purification of monocytes is dispensable prior to experiments requiring adherent monocytes as a starting material.