Dulbecco's phosphate-buffered saline (DPBS)
Lieferant: PROMOCELL
C-40230
C-40230EA
30
EUR
C-40230
Dulbecco's phosphate-buffered saline (DPBS)
Puffer
Gewebekultur-Puffer
Dulbecco's PBS solution for use in tissue culture, cell isolation procedures and molecular biology.
- Ready to use buffers
- Storage at 4 to 8 °C
Routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level.