Dulbecco's phosphate-buffered saline (DPBS)
Lieferant: PROMOCELL
C-40230
C-40230EA 30 EUR
C-40230
Dulbecco's phosphate-buffered saline (DPBS)
Puffer Gewebekultur-Puffer
Dulbecco's PBS solution for use in tissue culture, cell isolation procedures and molecular biology.
  • Ready to use buffers
  • Storage at 4 to 8 °C
Routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level.