Dulbecco's phosphate-buffered saline (DPBS) without Ca²⁺ and Mg²⁺
Lieferant: PROMOCELL
These dulbecco's PBS solution are used in tissue culture, cell isolation procedures and molecular biology.
- Ready to use buffers
- Storage at 4 to 8 °C
Routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level.