Trypsin neutralizing solution
Lieferant: PROMOCELL
Trypsin is the most widely employed enzyme used for cell detachment. This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0,05% trypsin inhibitor in 0,1% BSA.
Bitte beachten For in vitro research use only and not for diagnostic or therapeutic procedures.