Human accutase solution
Lieferant: PROMOCELL
C-41310
C-41310EA 71 EUR
C-41310
Human accutase solution
Enzyme
Accutase is a sterile filtered solution specially developed for gentle and effective cell dissociation.
  • Ready to use
  • No stabilisers or other proteins have been added
  • Accutase supplied in Dulbecco's PBS containing 0,5 mM EDTA
Accutase solution is a unique mixture of proteolytic and collagenolytic enzymes from an invertebrate species. Accutase has been ultra-filtered before lyophilisation and contains only purified enzymes.