Detachment reagents, macrophage detachment solution DXF
Lieferant: PROMOCELL
C-41330
C-41330EA 105 EUR
C-41330
Detachment reagents, macrophage detachment solution DXF
Zellkultur-Ergänzungen und -Zusätze
Macrophage Detachment Solution DXF is a non-enzymatic reagent especially optimised for the gentle detachment of strongly adherent macrophages.