Crystal ponceau 6R, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

C0644-5G
C0644-5GEA 154 EUR
C0644-5G
Crystal ponceau 6R, Sigma-Aldrich®
Crystal ponceau 6R

Crystal Ponceau 6R has been used in the MSB (martius yellow–crystal scarlet–methyl blue) staining procedure for the staining of collagen fibers and erythrocytes.

Formel: C₂₀H₁₂N₂Na₂O₇S₂
Schmelzpunkt: >300 °C (Lit.)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2766-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95243f5280c4ad4bb75a0cb8327fe7dadcee1f5e&vid=aa54253b600dcdf1eb55a61e5b108fa4066ccc6c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR