Carmin (Calcium-Aluminium-Verlackung von Carminsäure), powder, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: CI 75470

MQ200

C1022-100G C1022-25G C1022-5G
C1022-100GEA 890 EUR
C1022-100G C1022-25G C1022-5G
Carmin (Calcium-Aluminium-Verlackung von Carminsäure), powder, Sigma-Aldrich®
Carmin (Calcium-Aluminium-Verlackung von Carminsäure)

Carmine is a red compound and is generally used with aluminum, iron and other metal salts to enhance its activity. During smear preparation, it is required for chromatin staining in fresh material. In presence of aluminum chloride, it is used for the staining of glycogen.


  • Carmine has been used for the staining of mammary glands
  • It has been used for the staining of the chromosomes

Formel: C₂₂H₂₀O₁₃
Molecular Weight: 492.389 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00167028
CAS-Nummer: 1390-65-4
EINECS: 215-724-4

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Red
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour)
1 mg/ml in 2 N NH₄OH
Red
Assay (UV-Vis)
% Carminic acid
≥42.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR