4-Chloro-7-sulphobenzofurazan ammonium salt, Pulver

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 7-Chloro-2,1,3-benzoxadiazole-4-sulphonic acid ammonium salt, SBF-Chloride, 7-Chloro-4-benzofurazansulphonic acid ammonium salt, 4-Chloro-7-sulphobenz-2-oxa-1,3-diazole ammonium salt

M-Clarity™ Qualitätsstufe = MQ200

C1154-100MG
C1154-100MGEA 319 EUR
C1154-100MG
4-Chloro-7-sulphobenzofurazan ammonium salt, Pulver
4-Chloro-7-sulphobenzofurazan ammonium salt
Formel: C₆H₃ClN₂O₄S·NH₃
Molecular Weight: 251,65 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Room Temperature
MDL: MFCD00038995
CAS-Nummer: 81377-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e21d1472d725a3b80cc4214670d54ec3fe41117e&vid=e21d1472d725a3b80cc4214670d54ec3fe41117e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR