7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 1H-Pyrrole-2,5-dione, 1-(4-(7-(diethylamino)-4-methyl-2-oxo-2H-1-benzopyran-3-yl)phenyl)-, CPM

C1484-25MG
C1484-25MGEA 530 EUR
C1484-25MG
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM), Sigma-Aldrich®
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM)

7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin has been used: as a nontoxic substitute for conjugation with antibody(93) to aid fluorescent detection of Coenzyme A (CoA-SH) in human N-myristoyltransferases assay (94)

Formel: C₂₄H₂₂N₂O₄
Molecular Weight: 402.44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077334
CAS-Nummer: 76877-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6044c5c2f858f50ca881b51adf6f8ced10ef5907&vid=f4e665018ad48c82ff22573b93a0b8686ec8b494&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR