Calmagit Indikator-Qualität, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
C204-10G-A
C204-25G-A
C204-10G-AEA
123
EUR
C204-10G-A
C204-25G-A
Calmagit Indikator-Qualität, Supelco®
Calmagit
Used as an indicator in the titration of calcium or magnesium with EDTA. Used for the determination of magnesium in biological materials.
Formel:
C₁₇H₁₄N₂O₅S Molecular Weight: 358.37 g/mol |
MDL:
MFCD00011656 CAS-Nummer: 3147-14-6 EINECS: 221-563-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...