5-Carboxytetramethylrhodamine (5-TAMRA), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer
C2734-10MG
C2734-10MGEA
569
EUR
C2734-10MG
5-Carboxytetramethylrhodamine (5-TAMRA), Sigma-Aldrich®
5-Carboxytetramethylrhodamine (5-TAMRA)
An important fluorophore prepared in high purtity with a detection limit of TMR-labeled amino acids by capillary electrophoresis of approx. 600 molecules. Readily excited by the 546 nm spectral line from mercury-arc lamps used in fluorescent microscopes, and is intrinsically more photostable than fluorescein.
Formel:
C₂₅H₂₂N₂O₅ Molecular Weight: 430,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269769 CAS-Nummer: 91809-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...