Essigsäure (Eisessig) 99,5-100,5% USP, FCC, ACS, Macron Fine Chemicals™
Lieferant: Avantor
|
|
C3121-04EA
63.45
EUR
C3121-04
3121-46
Essigsäure (Eisessig) 99,5-100,5% USP, FCC, ACS, Macron Fine Chemicals™
Essigsäure
Bitte beachten Preserve in tight containers, and store at room temperature
Formel:
H₃CCOOH Siedepunkt: 118 °C (1013 hPa) Schmelzpunkt: 17 °C Dichte: 1,05 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036152 CAS-Nummer: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8,II |
|
Spezifikation Testergebnisse
Meets U.S.P Requirements | — |
Meets ACS Reagent Chemical Requirements | — |
Meets F.C.C. Requirements | — |
GMP Manufactured Product | — |
Food GMP Manufactured Product | — |
IMPORTANT: Material will freeze if stored below 17 °C (63°F). | — |
CAUTION: For Manufacturing, processing or repackaging | — |
Bulk Food Chemical | — |
Bulk Pharmaceutical Chemical | — |
USP - Assay (CH₃COOH) | 99.5 - 100.5 % |
USP - Chloride (Cl) (USP) | Passes Test |
USP - Congealing Temperature | ≥15.6 °C |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Readily Oxidizable Substances | Passes Test |
USP - Sulfate (USP)(SO₄) | Passes Test |
USP - Limit of Nonvolatile Residue | ≤0.005 % |
FCC - Assay (CH₃COOH) | 99.7 - 100.5 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Lead (Pb) | ≤0.5 mg/kg |
FCC - Limit of Nonvolatile Residue | ≤0.005 % |
FCC - Readily Oxidizable Substances | Passes Test |
FCC - Solidification Point | ≥15.6 °C |
ACS - Assay (by GC, corrected for water) | ≥99.7 % |
ACS - Acetic Anhydride ((CH₃CO)₂O) | ≤0.01 % |
ACS - Chloride (Cl) | ≤1 ppm |
ACS - Color (APHA) | ≤10 |
Dilution Test | Passes Test |
Trace Impurities - ACS - Iron (Fe) | ≤0.200 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤0.5 ppm |
ACS - Substances Reducing Dichromate | Passes Test |
ACS - Substances Reducing Permanganate | Passes Test |
ACS - Sulfate (SO₄) | ≤1 ppm |
ACS - Titrable Base (meq/g) | ≤0.0004 |
Copper (Cu) | ≤0.00001 % |
Nickel (Ni) | ≤0.00001 % |
Residue after Evaporation | ≤0.001 % |
Appearance (clear, colorless liquid) | — |
Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or | |
produced by the production process. | |
Preserve in tight containers, and store at room temperature | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...