Dextrose, wasserfrei 97,5-102,0% (durch Trockensubstanz), körnig USP, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Dextrose, Traubenzucker

C4908-88EA 549 EUR
C4908-88
Dextrose, wasserfrei 97,5-102,0% (durch Trockensubstanz), körnig USP, J.T.Baker®
D(+)-Glucose

These products are a reliable source of carbon and energy for cell cultures, and used as excipients, effectively stabilise valuable and delicate lyophilised proteins for injectable biopharmaceutical drugs.

Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 146 °C
Dichte: 1,54 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product Passes Test
Meets U.S.P Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging ≤1.0%
Bulk Pharmaceutical Chemical 97.5 - 102.0%
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
USP - Assay (Anhydrous basis) 97.5 - 102.0 %
USP - Related substances: Maltose and isomaltose ≤0.4 %
USP - Related substances: Maltotriose ≤0.2 %
USP - Related substances: Fructose ≤0.15 %
USP - Related substances: Unspecified impurities, each ≤0.10 %
USP - Related substances: Total Impurities ≤0.5 %
USP - Color and Clarity of Solution Passes Test
USP - Soluble Starch, Sulfites ≤15 ppm
USP - Conductivity (uS cm-1) ≤20 µS/cm
USP - Dextrin Passes Test
USP - Endotoxin Concentration (EU/g) ≤2.5
Solubility Passes Test
Appearance (white, granular powder)
Odorless
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR