5-CFDA (5-Carboxyfluorescein diacetate), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 5-Carboxy-di-O-acetylfluorescein, 5-CFDA, 5-Carboxyfluorescein diacetate

C4916-100MG
C4916-100MGEA 538 EUR
C4916-100MG
5-CFDA (5-Carboxyfluorescein diacetate), Sigma-Aldrich®
5-CFDA (5-Carboxyfluorescein diacetate)

5-Carboxyfluorescein diacetate has been used for the staining of bacteria to study bacterial adhesion to mucin.

Formel: C₂₅H₁₆O₉
Lagertemperatur: Gefrierschrank
MDL: MFCD00036872
CAS-Nummer: 79955-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90f25fab04d814422b457013bcf0d765cb125546&vid=bf98654826df16bd17c6d4790c746c2ba5b3a462&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR