Calcimycin (mixed Mg/CA salt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: A23187

C5149-5MG
C5149-5MGEA 209 EUR
C5149-5MG
Calcimycin (mixed Mg/CA salt), Sigma-Aldrich®
Calcimycin (mixed Mg/CA salt)

Ionophore highly selective for Ca2+. Potentiates responses to NMDA but not to quisqualate. In cell culture, stimulates nitric oxide production by calmodulin-dependent constitutive nitric oxide synthase.

Formel: (C₂₉H₃₆N₃O₆)₂·Mg(C₂₉H₃₆N₃O₆)₂·Ca
Lagertemperatur: Kühlschrank
CAS-Nummer: 52665-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1ce0b3fab00bb543b9b1f32c9a1c4617b350375&vid=1f35d0b3a4badb4fe64ffe5f9501bacfe296702d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR